Learn More
Medchemexpress LLC Pitavastatin-d4 | 2070009-71-9 | 98.8% | 5 MG
SDP
Catalog No. 5000450911
Encompass_Preferred
Pitavastatin-d4 is a deuterium-labeled form of Pitavastatin, a potent HMG-CoA reductase inhibitor. It is effective in inhibiting cholesterol synthesis and acts as an efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer.
  • Deuterium-labeled compound
  • Potent HMG-CoA reductase inhibitor
  • Inhibits cholesterol synthesis from acetic acid
  • Efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer
  • Possesses anti-atherosclerotic effects
  • Possesses anti-asthmatic and anti-osteoarthritis effects
  • Exhibits antineoplastic, neuroprotective, hepatoprotective, and reno-protective effects
Catalog No. 50-004-50911 Supplier Medchemexpress LLC Supplier No. HYB0144AS5MG
Pitavastatin-d4 is a deuterium-labeled form of Pitavastatin, a potent HMG-CoA reductase inhibitor. It is effective in inhibiting cholesterol synthesis and acts as an efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer.
  • Deuterium-labeled compound
  • Potent HMG-CoA reductase inhibitor
  • Inhibits cholesterol synthesis from acetic acid
  • Efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer
  • Possesses anti-atherosclerotic effects
  • Possesses anti-asthmatic and anti-osteoarthritis effects
  • Exhibits antineoplastic, neuroprotective, hepatoprotective, and reno-protective effects