Learn More
Medchemexpress LLC Pitavastatin-d4 sodium | 574705-92-3 | 99.7% | 1 MG
SDP
Catalog No. 5000450912
Encompass_Preferred
Pitavastatin-d4 sodium is a deuterium-labeled version of Pitavastatin sodium. Deuteration, the process of incorporating stable heavy isotopes of hydrogen (deuterium) into drug molecules, has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50912 Supplier Medchemexpress LLC Supplier No. HYB0144BS1MG
Add to Cart
Add to Cart
Pitavastatin-d4 sodium is a deuterium-labeled version of Pitavastatin sodium. Deuteration, the process of incorporating stable heavy isotopes of hydrogen (deuterium) into drug molecules, has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS