Learn More
Medchemexpress LLC Pitavastatin-d4 sodium | 99.7% | 100 UG
SDP
Supplier:  Medchemexpress LLC HYB0144BS100UG
Encompass_Preferred
Pitavastatin-d4 sodium is a deuterium-labeled version of Pitavastatin sodium. It is intended for research use only and can be utilized as a tracer or as an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets Autophagy, Apoptosis, Mitophagy, and HMG-CoA Reductase (HMGCR)
  • Belongs to Isotope-Labeled Compounds
Catalog No. 50-004-50773