Learn More
Medchemexpress LLC Pitavastatin-d5 sodium | 574705-92-3 | 99.5% | 1 MG
SDP
Catalog No. 5000450475
Encompass_Preferred
Pitavastatin-d5 sodium is the deuterium-labeled Pitavastatin sodium. Pitavastatin (NK-104) is a potent hydroxymethylglutaryl-CoA (HMG-CoA) reductase inhibitor, inhibiting cholesterol synthesis with an IC50 of 5.8 nM in HepG2 cells. It also acts as an efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer and exhibits anti-cancer activity.
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent HMG-CoA reductase inhibitor
  • Inhibits cholesterol synthesis from acetic acid
  • Efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer
  • Exhibits anti-cancer activity
Catalog No. 50-004-50475 Supplier Medchemexpress LLC Supplier No. HYB0144AS11MG
Pitavastatin-d5 sodium is the deuterium-labeled Pitavastatin sodium. Pitavastatin (NK-104) is a potent hydroxymethylglutaryl-CoA (HMG-CoA) reductase inhibitor, inhibiting cholesterol synthesis with an IC50 of 5.8 nM in HepG2 cells. It also acts as an efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer and exhibits anti-cancer activity.
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent HMG-CoA reductase inhibitor
  • Inhibits cholesterol synthesis from acetic acid
  • Efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer
  • Exhibits anti-cancer activity