Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pitavastatin lactone | 141750-63-2 | 99.3% | 403.45 g/mol | C25H22FNO3 | 10MG
SDP

Catalog No. 5000235282
Encompass_Preferred

Pitavastatin lactone is the lactone metabolite of pitavastatin and is supplied as a research-grade reference compound for metabolite identification, analytical method development, and pharmacokinetic studies. The material is provided as a high-purity solid with specified storage recommendations for stability.

  • High purity: 99.3%.
  • Molecular weight: 403.45 g/mol.
  • Chemical formula: C25H22FNO3.
  • Appearance: white to off-white solid.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Common applications: metabolite standard, LC-MS method validation, pharmacokinetic studies.

Catalog No. 50-002-35282 Supplier Medchemexpress LLC Supplier No. HY12564310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitavastatin lactone is the lactone metabolite of pitavastatin and is supplied as a research-grade reference compound for metabolite identification, analytical method development, and pharmacokinetic studies. The material is provided as a high-purity solid with specified storage recommendations for stability.

  • High purity: 99.3%.
  • Molecular weight: 403.45 g/mol.
  • Chemical formula: C25H22FNO3.
  • Appearance: white to off-white solid.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Common applications: metabolite standard, LC-MS method validation, pharmacokinetic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.