Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pitavastatin lactone | 141750-63-2 | 99.3% | 403.45 g/mol | C25H22FNO3 | 25MG
SDP

Supplier:  Medchemexpress LLC HY12564325MG

Encompass_Preferred

Pitavastatin lactone is the lactone metabolite of pitavastatin, provided as a high-purity analytical standard for in vitro and metabolic studies. The material is supplied as a white to off-white solid with specified solubility in DMSO and recommended low-temperature storage to preserve stability and integrity for analytical workflows.

  • High purity: 99.3%.
  • Molecular formula: C25H22FNO3; molecular weight: 403.45 g/mol.
  • Appearance: white to off-white solid.
  • Solubility: DMSO 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder at -20°C (stable up to 3 years); in solvent -80°C for 6 months or -20°C for 1 month.
  • Supplied in small pack sizes suitable for analytical testing.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-36068


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.