Learn More
Medchemexpress LLC Pitavastatin lactone | 141750-63-2 | MFCD09841193 | 99.3% | 403.45 g/mol | C25H22FNO3 | 50MG
SDP
Catalog No. 5000238128
Encompass_Preferred
Pitavastatin lactone is the lactone metabolite of pitavastatin provided as a high-purity reference standard for research and analytical applications. It is intended for use in metabolic and pharmacokinetic studies, impurity profiling, and chromatographic method development (research use only).
  • High purity suitable for analytical reference (99.3% by HPLC).
  • Confirmed chemical identity and formula: C25H22FNO3, MW 403.45 g/mol.
  • Supplied in small laboratory quantities for research use.
  • Suitable for metabolic and pharmacokinetic studies, and method development.
  • Provided with supporting documentation such as datasheet, COA, and SDS.
Catalog No. 50-002-38128 Supplier Medchemexpress LLC Supplier No. HY12564350MG
Pitavastatin lactone is the lactone metabolite of pitavastatin provided as a high-purity reference standard for research and analytical applications. It is intended for use in metabolic and pharmacokinetic studies, impurity profiling, and chromatographic method development (research use only).
  • High purity suitable for analytical reference (99.3% by HPLC).
  • Confirmed chemical identity and formula: C25H22FNO3, MW 403.45 g/mol.
  • Supplied in small laboratory quantities for research use.
  • Suitable for metabolic and pharmacokinetic studies, and method development.
  • Provided with supporting documentation such as datasheet, COA, and SDS.