Learn More
Cayman Chemical Pitavastatin (calcium salt), Purity: ≥98%, CAS: 147526-32-7, 10 mg
SDP
Catalog No. 502969383
Encompass_Preferred
An HMG-CoA reductase inhibitor (Ki = 1.7 nM for the rat liver microsomal enzyme); inhibits sterol biosynthesis in rat liver and ileum (ED50s = 0.13 and 0.2 mg/kg, respectively); reduces plasma levels of triglycerides and total cholesterol in dogs; increases survival in Dahl salt-sensitive rats fed a high-salt diet at 0.3 mg/kg
Catalog No. 50-296-9383 Supplier Cayman Chemical Supplier No. 1541410 MG
An HMG-CoA reductase inhibitor (Ki = 1.7 nM for the rat liver microsomal enzyme); inhibits sterol biosynthesis in rat liver and ileum (ED50s = 0.13 and 0.2 mg/kg, respectively); reduces plasma levels of triglycerides and total cholesterol in dogs; increases survival in Dahl salt-sensitive rats fed a high-salt diet at 0.3 mg/kg