Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Reagent, PITC, 1 mL, Solid, Reversed Phase

Catalog No. 50466777 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This PITC (phenylisothiocyanate) reagent is a precolumn derivatization reagent used in the PicoTag Method for amino acid analysis. It is designed to reveal structural information about amino acids in a polypeptide chain.

  • Precolumn derivatization for amino acid analysis
  • Separation mode: reversed phase
  • Detection method: UV
  • Formulation: solid
  • Storage condition: room temperature
  • System type: LC
  • Shipping condition: ambient
  • Volume: 1 mL
  • Application: protein digestion
  • Units per package: 10 pk
  • Contains hazardous material requiring special freight handling and additional charges

Catalog No. 50-466-777 Supplier Waters Corp Supplier No. WAT088120
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This PITC (phenylisothiocyanate) reagent is a precolumn derivatization reagent used in the PicoTag Method for amino acid analysis. It is designed to reveal structural information about amino acids in a polypeptide chain.

  • Precolumn derivatization for amino acid analysis
  • Separation mode: reversed phase
  • Detection method: UV
  • Formulation: solid
  • Storage condition: room temperature
  • System type: LC
  • Shipping condition: ambient
  • Volume: 1 mL
  • Application: protein digestion
  • Units per package: 10 pk
  • Contains hazardous material requiring special freight handling and additional charges

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.