Learn More
Bellco Glass, Inc. Pitch Blade Assembly , 7L
SDP
Catalog No. 501944736
Encompass_Preferred
Stainless steel pitch blade impeller 7l MADE IN THE USA 316l stainless steel Ra 32 finish or better
Catalog No. 50-194-4736 Supplier Bellco Glass, Inc. Supplier No. 196402310
Stainless steel pitch blade impeller 7l MADE IN THE USA 316l stainless steel Ra 32 finish or better