Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO PITCH BLADE DOWN 30MM-HC-4X IM

Catalog No. 502741804 Shop All METTLER TOLEDO Products
Encompass

Pitch Blade Down 30mm-HC-4x Impeller

Catalog No. 50-274-1804 Supplier METTLER TOLEDO Supplier No. 30030951
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitch Blade Down 30mm-HC-4x Impeller

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.