Learn More
Sklar Instruments Pitcher/Beaker, Sklar, Stainless Steel.This product is 3-3/8 in. x 4-5/8 in. with a volume of 600ml. It does not have a handle. EA/1.
SDP
Catalog No. 501436938 Shop All Sklar Instruments Products
Encompass_Preferred
Pitcher/Beaker, Sklar, Stainless Steel. Sklar's Stainless Steel Beaker may be used for a variety of things, including use in the laboratory, or holding sterilized instruments in solution until their use in specific procedures. This product is 3-3/8 in. x 4-5/8 in. with a volume of 600ml. It does not have a handle.
This item is not returnable. View return policy
Catalog No. 50-143-6938 Supplier Sklar Instruments Supplier No. 101174
Pitcher/Beaker, Sklar, Stainless Steel. Sklar's Stainless Steel Beaker may be used for a variety of things, including use in the laboratory, or holding sterilized instruments in solution until their use in specific procedures. This product is 3-3/8 in. x 4-5/8 in. with a volume of 600ml. It does not have a handle.