Learn More
Sklar Instruments Pitcher/Beaker, Sklar, Stainless Steel. This product is 4-7/8 in. x 7-1/8 in. with a volume of 2000ml. It does not have a handle. EA/1.
SDP
Catalog No. 501436940 Shop All Sklar Instruments Products
Encompass_Preferred
Pitcher/Beaker, Sklar, Stainless Steel. Sklar's Stainless Steel Beaker may be used for a variety of things, including use in the laboratory, or holding sterilized instruments in solution until their use in specific procedures. This product is 4-7/8 in. x 7-1/8 in. with a volume of 2000ml. It does not have a handle.
This item is not returnable. View return policy
Catalog No. 50-143-6940 Supplier Sklar Instruments Supplier No. 101176
Pitcher/Beaker, Sklar, Stainless Steel. Sklar's Stainless Steel Beaker may be used for a variety of things, including use in the laboratory, or holding sterilized instruments in solution until their use in specific procedures. This product is 4-7/8 in. x 7-1/8 in. with a volume of 2000ml. It does not have a handle.