Learn More
Sklar Instruments Pitcher/Beaker, Sklar, Stainless Steel. This product is 5-1/16 in. x 6-5/8 in. with a volume of 2000ml. EA/1.
SDP
Catalog No. 501437040 Shop All Sklar Instruments Products
Encompass_Preferred
Pitcher/Beaker, Sklar, Stainless Steel. Sklar's Stainless Steel Pitcher / Beaker may be used for a variety of things, including use in the laboratory, or holding sterilized instruments in solution until their use in specific procedures. This product is 5-1/16 in. x 6-5/8 in. with a volume of 2000ml. It also has a handle.
This item is not returnable. View return policy
Catalog No. 50-143-7040 Supplier Sklar Instruments Supplier No. 101539
Pitcher/Beaker, Sklar, Stainless Steel. Sklar's Stainless Steel Pitcher / Beaker may be used for a variety of things, including use in the laboratory, or holding sterilized instruments in solution until their use in specific procedures. This product is 5-1/16 in. x 6-5/8 in. with a volume of 2000ml. It also has a handle.