Learn More
Hach Company Pitcher, Graduated, 1000 cc
Supplier: Hach Company 2612853
Chemical-resistant, shatter-proof, tall-form pitchers with well-designed spouts, and large handles. Metric graduations are molded onto outside wall of each pitcher. Polypropylene, steam autoclavable at 250°F (121°C).