Learn More
Hach Company Pitcher, Graduated, 2000 cc
Catalog No. 501996348 Shop All Hach Company Products
Encompass
Chemical-resistant, shatter-proof, tall-form pitchers with well-designed spouts, and large handles. Metric graduations are molded onto outside wall of each pitcher. Polypropylene, steam autoclavable at 250°F (121°C).
Catalog No. 50-199-6348 Supplier Hach Company Supplier No. 2612854
Chemical-resistant, shatter-proof, tall-form pitchers with well-designed spouts, and large handles. Metric graduations are molded onto outside wall of each pitcher. Polypropylene, steam autoclavable at 250°F (121°C).