Learn More
Pith Balls
SCI_ED
Catalog No. p-2826976
Change view
Click to view available options
Includes:
Without string
Material:
Aluminum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Material
S65015 Without string Aluminum
S65014 Without string -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S65015 Supplier Lab Connections Supplier No. 15033
For electrostatic demonstrations.
  • Available with or without strings
  • Ultra-light and ultra-sensitive
Specifications
Material Aluminum
For Use With (Application) Static electricity, polarity, friction studies
Product Type Pith Ball
Includes Without string
Size 1 x 1cm; 6/Bag
Dimensions (L x W) 1 x 1 cm; 6/Bag