Learn More
Medchemexpress LLC HY-110389 5mg Medchemexpress, Pitofenone hydrochloride CAS:1248-42-6 Purity:>98%
SDP
Catalog No. 501968795
Encompass_Preferred
Medchemexpress, HY-110389 5mg Pitofenone HCl CAS:1248-42-6 Pitofenone hydrochloride, a spasmolytic compound, inhibits the acetylcholinesterase (AChE) activity from bovine erythrocytes and from electric eel with Kis of 36 and 45 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8795 Supplier Medchemexpress LLC Supplier No. HY1103895MG
Medchemexpress, HY-110389 5mg Pitofenone HCl CAS:1248-42-6 Pitofenone hydrochloride, a spasmolytic compound, inhibits the acetylcholinesterase (AChE) activity from bovine erythrocytes and from electric eel with Kis of 36 and 45 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.