Learn More
Cayman Chemical PItofenonhydrochlorIde 10mg
SDP
Catalog No. 502995099
Encompass_Preferred
An antispasmodic agent; an inhibitor of AChE (Kis = 36 and 45 μM for bovine erythrocyte and electric eel enzyme, respectively); inhibits acetylcholine-induced contractions of isolated guinea pig ileum at 2.5 μM
Catalog No. 50-299-5099 Supplier Cayman Chemical Supplier No. 2386510 MG
An antispasmodic agent; an inhibitor of AChE (Kis = 36 and 45 μM for bovine erythrocyte and electric eel enzyme, respectively); inhibits acetylcholine-induced contractions of isolated guinea pig ileum at 2.5 μM