Learn More
Cayman Chemical Pitofenone (hydrochloride), Purity: ≥98%, CAS: 1248-42-6, 50 mg
SDP
Catalog No. 502995100
Encompass_Preferred
An antispasmodic agent; an inhibitor of AChE (Kis = 36 and 45 μM for bovine erythrocyte and electric eel enzyme, respectively); inhibits acetylcholine-induced contractions of isolated guinea pig ileum at 2.5 μM
Catalog No. 50-299-5100 Supplier Cayman Chemical Supplier No. 2386550 MG
An antispasmodic agent; an inhibitor of AChE (Kis = 36 and 45 μM for bovine erythrocyte and electric eel enzyme, respectively); inhibits acetylcholine-induced contractions of isolated guinea pig ileum at 2.5 μM