Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pitolisant | 362665-56-3 | 99.7% | 25 MG
SDP

Catalog No. 5000231611
Encompass_Preferred

Pitolisant is a potent, selective nonimidazole inverse agonist of the human histamine H3 receptor (Ki = 0.16 nM). Supplied as an analytical research standard in a 25 mg quantity, it is used in receptor pharmacology, biochemical assays, and analytical method development.

  • Potent H3 receptor inverse agonist with Ki = 0.16 nM.
  • High purity analytical standard suitable for assay calibration.
  • Offered in a 25 mg quantity appropriate for laboratory studies.
  • Chemical formula C17H26ClNO; molecular weight 295.85 g·mol⁻¹.
  • Recommended storage: pure form at -20°C for long-term stability.

Catalog No. 50-002-31611 Supplier Medchemexpress LLC Supplier No. HY1219925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitolisant is a potent, selective nonimidazole inverse agonist of the human histamine H3 receptor (Ki = 0.16 nM). Supplied as an analytical research standard in a 25 mg quantity, it is used in receptor pharmacology, biochemical assays, and analytical method development.

  • Potent H3 receptor inverse agonist with Ki = 0.16 nM.
  • High purity analytical standard suitable for assay calibration.
  • Offered in a 25 mg quantity appropriate for laboratory studies.
  • Chemical formula C17H26ClNO; molecular weight 295.85 g·mol⁻¹.
  • Recommended storage: pure form at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.