Learn More
Selleck Chemical LLC Pitolisant hydrochloride
SDP
Catalog No. 5000003932
Encompass_Preferred
Pitolisant (Tiprolisant BF-2649) acts as a high-affinity competitive antagonist (Ki 0 16 nM) and as an inverse agonist (EC50 1 5 nM) at the human histamine H3 receptor subtype
Catalog No. 50-000-03932 Supplier Selleck Chemical LLC Supplier No. S592625MG
Pitolisant (Tiprolisant BF-2649) acts as a high-affinity competitive antagonist (Ki 0 16 nM) and as an inverse agonist (EC50 1 5 nM) at the human histamine H3 receptor subtype