Learn More
Medchemexpress LLC Pitolisant hydrochloride | 903576-44-3 | 5 MG
SDP
Catalog No. 5000230446
Encompass_Preferred
Pitolisant hydrochloride is the hydrochloride salt of pitolisant, a potent and selective non-imidazole inverse agonist of the human histamine H3 receptor used in pharmacology and neuroscience research. It is supplied as a solid and is commonly prepared as a solution for in vitro assays or dosed in vivo for preclinical studies.
  • Potent H3 receptor inverse agonist (reported Ki = 0.16 nM).
  • Hydrochloride salt; molecular weight 332.31 g/mol.
  • Typical research applications include histaminergic signaling and sleep disorder models.
  • Supplied in small milligram pack sizes for research use.
  • CAS 903576-44-3 for unambiguous identification.
Catalog No. 50-002-30446 Supplier Medchemexpress LLC Supplier No. HY12199B5MG
Pitolisant hydrochloride is the hydrochloride salt of pitolisant, a potent and selective non-imidazole inverse agonist of the human histamine H3 receptor used in pharmacology and neuroscience research. It is supplied as a solid and is commonly prepared as a solution for in vitro assays or dosed in vivo for preclinical studies.
  • Potent H3 receptor inverse agonist (reported Ki = 0.16 nM).
  • Hydrochloride salt; molecular weight 332.31 g/mol.
  • Typical research applications include histaminergic signaling and sleep disorder models.
  • Supplied in small milligram pack sizes for research use.
  • CAS 903576-44-3 for unambiguous identification.