Learn More
Medchemexpress LLC Pitolisant hydrochloride | 903576-44-3 | ≥98.0% | 10 MG
SDP
Catalog No. 5000231154
Encompass_Preferred
Pitolisant hydrochloride is a potent, selective nonimidazole inverse agonist of the human histamine H3 receptor supplied as a research-grade hydrochloride salt for laboratory use.
  • High potency (Ki = 0.16 nM).
  • Research reagent for in vitro pharmacology and receptor studies.
  • White to beige powder, supplied in small quantities for laboratory use (e.g., 10 MG).
  • Purity ≥98.0% (HPLC) as commonly reported for research-grade material.
  • Stable when stored sealed and away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-31154 Supplier Medchemexpress LLC Supplier No. HY12199B10MG
Pitolisant hydrochloride is a potent, selective nonimidazole inverse agonist of the human histamine H3 receptor supplied as a research-grade hydrochloride salt for laboratory use.
  • High potency (Ki = 0.16 nM).
  • Research reagent for in vitro pharmacology and receptor studies.
  • White to beige powder, supplied in small quantities for laboratory use (e.g., 10 MG).
  • Purity ≥98.0% (HPLC) as commonly reported for research-grade material.
  • Stable when stored sealed and away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).