Learn More
Medchemexpress LLC Pitolisant hydrochloride | 903576-44-3 | 99.5% | 1 ML
SDP
Catalog No. 5000230326
Encompass_Preferred
Pitolisant hydrochloride is a research-grade histamine H3 receptor inverse agonist/antagonist supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research use. CAS: 903576-44-3.
Catalog No. 50-002-30326 Supplier Medchemexpress LLC Supplier No. HY12199B10MM 1ML
Pitolisant hydrochloride is a research-grade histamine H3 receptor inverse agonist/antagonist supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research use. CAS: 903576-44-3.