Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pitolisant hydrochloride | 903576-44-3 | 99.5% | 100 MG
SDP

Catalog No. 5000231561
Encompass_Preferred

Pitolisant hydrochloride is the hydrochloride salt of pitolisant, a potent and selective non-imidazole inverse agonist at the human histamine H3 receptor (Ki = 0.16 nM). It is supplied as a research-grade chemical with documented solubility and storage recommendations for in vitro and in vivo use.

  • Potent H3 receptor inverse agonist (Ki = 0.16 nM).
  • Chemical formula C17H27Cl2NO; molecular weight 332.31 g/mol.
  • High purity: 99.5%.
  • Soluble in water and DMSO; specific solubility data available for in vivo formulations.
  • Store sealed away from moisture; stable in solution at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-31561 Supplier Medchemexpress LLC Supplier No. HY12199B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitolisant hydrochloride is the hydrochloride salt of pitolisant, a potent and selective non-imidazole inverse agonist at the human histamine H3 receptor (Ki = 0.16 nM). It is supplied as a research-grade chemical with documented solubility and storage recommendations for in vitro and in vivo use.

  • Potent H3 receptor inverse agonist (Ki = 0.16 nM).
  • Chemical formula C17H27Cl2NO; molecular weight 332.31 g/mol.
  • High purity: 99.5%.
  • Soluble in water and DMSO; specific solubility data available for in vivo formulations.
  • Store sealed away from moisture; stable in solution at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.