Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pitolisant hydrochloride | 903576-44-3 | 99.5% | 25 MG
SDP

Catalog No. 5000230881
Encompass_Preferred

Pitolisant hydrochloride is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki = 0.16 nM). It is supplied as the hydrochloride salt for research use only.

  • Potent inverse agonist at the histamine H3 receptor (Ki = 0.16 nM).
  • Hydrochloride salt form for research handling and stability.
  • Molecular weight 332.31 and formula C17H27Cl2NO.
  • CAS number 903576-44-3 for precise identification.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-30881 Supplier Medchemexpress LLC Supplier No. HY12199B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitolisant hydrochloride is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki = 0.16 nM). It is supplied as the hydrochloride salt for research use only.

  • Potent inverse agonist at the histamine H3 receptor (Ki = 0.16 nM).
  • Hydrochloride salt form for research handling and stability.
  • Molecular weight 332.31 and formula C17H27Cl2NO.
  • CAS number 903576-44-3 for precise identification.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.