Learn More
Aviva Systems Biology Corp PITPNM1 Antibody - N-terminal region (ARP59787_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59787_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PITPNM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PITPNM1. Alternate names: DRES9, FLJ44997, NIR2, PITPNM, RDGB, RDGB1, RDGBA, RDGBA1, Rd9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-824