Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pitstop 2 10mg | 1419320-73-2 | 10 MG
SDP

Catalog No. 5000229434
Encompass_Preferred

Pitstop 2 is a small-molecule clathrin inhibitor that binds the terminal domain of clathrin and blocks clathrin-mediated endocytosis. It is supplied for research use and has been used in studies of cell proliferation and cancer-related cellular processes.

  • Inhibits clathrin-mediated endocytosis.
  • Used in cell biology and anti-cancer research applications.
  • High purity (98.42%) as supplied.
  • Molecular weight 473.36 g/mol.
  • Soluble in DMSO at 50 mg/mL (105.63 mM); ultrasonic assistance recommended.
  • Store solid at 4°C protected from light; in solvent store at -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-29434 Supplier Medchemexpress LLC Supplier No. HY11560410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitstop 2 is a small-molecule clathrin inhibitor that binds the terminal domain of clathrin and blocks clathrin-mediated endocytosis. It is supplied for research use and has been used in studies of cell proliferation and cancer-related cellular processes.

  • Inhibits clathrin-mediated endocytosis.
  • Used in cell biology and anti-cancer research applications.
  • High purity (98.42%) as supplied.
  • Molecular weight 473.36 g/mol.
  • Soluble in DMSO at 50 mg/mL (105.63 mM); ultrasonic assistance recommended.
  • Store solid at 4°C protected from light; in solvent store at -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.