Learn More
Abcam Pitstop 2 - negative control, Negative control for clathrin inhibitor Pitstop2, 5MG
Catalog No. NC0282594
Encompass_Preferred
MW 409.3 g/mol, Purity >99%. Negative control for clathrin inhibitor Pitstop 2 (ab120687) which is of the same chemical class and has a highly related structure to Pitstop 2. Inhibits amphiphysin binding to the clathrin TD with an IC₅₀ >100 μM and does not block receptor-mediated endocytosis at concentrations of up to 300 μM (unpublished reports).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0282594 Supplier Abcam Supplier No. AB1206885MG
Add to Cart
Add to Cart
MW 409.3 g/mol, Purity >99%. Negative control for clathrin inhibitor Pitstop 2 (ab120687) which is of the same chemical class and has a highly related structure to Pitstop 2. Inhibits amphiphysin binding to the clathrin TD with an IC₅₀ >100 μM and does not block receptor-mediated endocytosis at concentrations of up to 300 μM (unpublished reports).
The product is subject to the following: Abcam Restricted Use Statement