Learn More
Medchemexpress LLC Pittsburgh compound B | 566169-93-5 | 98.0% | 256.33 g·mol⁻1 | C14H12N2OS | 5MG
SDP
Catalog No. 5000239569
Encompass_Preferred
Pittsburgh Compound B (PiB) is a research-grade PET tracer used to bind and visualize amyloid-β (Aβ) deposits for Alzheimer's disease research. Supplied as a small-molecule solid in milligram-scale quantities, it is intended for preclinical and in vitro studies, imaging assay development, and ligand-binding experiments.
  • High purity 98.0% for reproducible experimental results.
  • Supplied as 5 mg for small-scale assays and method development.
  • Binds amyloid-β enabling PET tracer and imaging applications.
  • Molecular formula C14H12N2OS and molecular weight 256.33 g·mol⁻¹ for precise stoichiometry.
  • Provided with product datasheet for handling and characterization details.
Catalog No. 50-002-39569 Supplier Medchemexpress LLC Supplier No. HY1261925MG
Pittsburgh Compound B (PiB) is a research-grade PET tracer used to bind and visualize amyloid-β (Aβ) deposits for Alzheimer's disease research. Supplied as a small-molecule solid in milligram-scale quantities, it is intended for preclinical and in vitro studies, imaging assay development, and ligand-binding experiments.
  • High purity 98.0% for reproducible experimental results.
  • Supplied as 5 mg for small-scale assays and method development.
  • Binds amyloid-β enabling PET tracer and imaging applications.
  • Molecular formula C14H12N2OS and molecular weight 256.33 g·mol⁻¹ for precise stoichiometry.
  • Provided with product datasheet for handling and characterization details.