Learn More
ABclonal Technology Pituitary homeobox 2 PITX2
SDP
Catalog No. 5000005123
Encompass_Preferred
This gene encodes a member of the RIEG PITX homeobox family which is in the bicoid class of homeodomain proteins The encoded protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression This protein plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes is involved in the development of the eye tooth and abdominal organs and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin Mutations in this gene are associated with Axenfeld-Rieger syndrome iridogoniodysgenesis syndrome and sporadic cases of Peters anomaly A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development Alternatively spliced transcript variants encoding distinct isoforms have been described
Catalog No. 50-000-05123 Supplier ABclonal Technology Supplier No. A2217320UL
This gene encodes a member of the RIEG PITX homeobox family which is in the bicoid class of homeodomain proteins The encoded protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression This protein plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes is involved in the development of the eye tooth and abdominal organs and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin Mutations in this gene are associated with Axenfeld-Rieger syndrome iridogoniodysgenesis syndrome and sporadic cases of Peters anomaly A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development Alternatively spliced transcript variants encoding distinct isoforms have been described