Learn More
Aviva Systems Biology Corp PITX2 Antibody - N-terminal region (ARP35634_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35634_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PITX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PITX2. Alternate names: RS, RGS, ARP1, Brx1, IDG2, IGDS, IHG2, PTX2, RIEG, IGDS2, IRID2, Otlx2, RIEG1ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55793