Learn More
Aviva Systems Biology Corp PITX2 Antibody - N-terminal region (ARP37981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PITX2. It was validated on Western Blot. . Alternate names: ARP1, Brx1, IDG2, IGDS, IGDS2, IHG2, IRID2, MGC111022, MGC20144, Otlx2, PTX2, RGS, RIEG, RIEG1, RSApplicationWB. Species reactivity: Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-705