Learn More
Medchemexpress LLC HY-108295 10mg Medchemexpress, Pivagabine CAS:69542-93-4 Purity:>98%
SDP
Catalog No. 501968520
Encompass_Preferred
Medchemexpress, HY-108295 10mg Pivagabine CAS:69542-93-4 Pivagabine (CXB 722) is a hydrophobic 4-aminobutyric acid derivative with neuromodulatory activity. Pivagabine penetrates the blood-brain barrier in rats. Pivagabine antagonizes the effects of foot shock on both GABAA receptor function and corticotropin-releasing factor (CRF) concentrations in rat brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8520 Supplier Medchemexpress LLC Supplier No. HY10829510MG
Medchemexpress, HY-108295 10mg Pivagabine CAS:69542-93-4 Pivagabine (CXB 722) is a hydrophobic 4-aminobutyric acid derivative with neuromodulatory activity. Pivagabine penetrates the blood-brain barrier in rats. Pivagabine antagonizes the effects of foot shock on both GABAA receptor function and corticotropin-releasing factor (CRF) concentrations in rat brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.