Learn More
Medchemexpress LLC HY-108295 5mg Medchemexpress, Pivagabine CAS:69542-93-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1082955MG
Encompass_Preferred
Medchemexpress, HY-108295 5mg Pivagabine CAS:69542-93-4 Pivagabine (CXB 722) is a hydrophobic 4-aminobutyric acid derivative with neuromodulatory activity. Pivagabine penetrates the blood-brain barrier in rats. Pivagabine antagonizes the effects of foot shock on both GABAA receptor function and corticotropin-releasing factor (CRF) concentrations in rat brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8521