Learn More
TARGETMOL CHEMICALS INC Pivmecillinam hydrochloride
SDP
Catalog No. 502877938
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active prodrug of mecillinam, an extended-spectrum penicillin antibiotic effective against gram-negative organisms, and is used as a substitute for amdinocillin. Purity 99.86%
Catalog No. 50-287-7938 Supplier TARGETMOL CHEMICALS INC Supplier No. T756050MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active prodrug of mecillinam, an extended-spectrum penicillin antibiotic effective against gram-negative organisms, and is used as a substitute for amdinocillin. Purity 99.86%