Learn More
Selleck Chemical LLC Pivmecillinam hydrochloride S5473-25mg
SDP
Catalog No. 503110254
Encompass_Preferred
Pivmecillinam is a -lactam antibiotic and a prodrug of mecillinam It has selective activity against Gram-negative bacteria and is used primarily in the treatment of lower urinary tract infections
Catalog No. 50-311-0254 Supplier Selleck Chemical LLC Supplier No. S547325MG
Pivmecillinam is a -lactam antibiotic and a prodrug of mecillinam It has selective activity against Gram-negative bacteria and is used primarily in the treatment of lower urinary tract infections