Learn More
Medchemexpress LLC Pivmecillinam hydrochloride | 32887-03-9 | 98.5% | 5 MG
SDP
Catalog No. 5000408088
Encompass_Preferred
Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active proagent of mecillinam, which is an extended-spectrum penicillin antibiotic. It acts as a bacterial inhibitor by targeting β-lactam. This compound has demonstrated effectiveness in inhibiting urinary tract infections caused by multidrug-resistant E. coli in animal models, with ongoing clinical trials for various related conditions.
  • Orally active proagent of mecillinam
  • Extended-spectrum penicillin antibiotic
  • Bacterial inhibitor targeting β-lactam
  • Effective against multidrug-resistant E. coli in UTIs in mouse models
  • Involved in clinical trials for pyelonephritis, UTIs, and bacteremia
Catalog No. 50-004-08088 Supplier Medchemexpress LLC Supplier No. HYB0810A5MG
Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active proagent of mecillinam, which is an extended-spectrum penicillin antibiotic. It acts as a bacterial inhibitor by targeting β-lactam. This compound has demonstrated effectiveness in inhibiting urinary tract infections caused by multidrug-resistant E. coli in animal models, with ongoing clinical trials for various related conditions.
  • Orally active proagent of mecillinam
  • Extended-spectrum penicillin antibiotic
  • Bacterial inhibitor targeting β-lactam
  • Effective against multidrug-resistant E. coli in UTIs in mouse models
  • Involved in clinical trials for pyelonephritis, UTIs, and bacteremia