Learn More
Medchemexpress LLC Pivmecillinam hydrochloride | 32887-03-9 | 99.19% | 1 ML
SDP
Catalog No. 5000408116
Encompass_Preferred
Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active proagent of mecillinam, which is an extended-spectrum penicillin antibiotic. It is intended for research use only and not for sale to patients. It appears as a white to off-white solid.
  • Orally active proagent of mecillinam
  • Extended-spectrum penicillin antibiotic
  • For research use only
  • White to off-white solid appearance
  • Available in 10 mM in 1 mL DMSO solution
Catalog No. 50-004-08116 Supplier Medchemexpress LLC Supplier No. HYB0810A10MM/1ML
Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active proagent of mecillinam, which is an extended-spectrum penicillin antibiotic. It is intended for research use only and not for sale to patients. It appears as a white to off-white solid.
  • Orally active proagent of mecillinam
  • Extended-spectrum penicillin antibiotic
  • For research use only
  • White to off-white solid appearance
  • Available in 10 mM in 1 mL DMSO solution