Learn More
Medchemexpress LLC Pivmecillinam hydrochloride | 32887-03-9 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0810AR5MG
Encompass_Preferred
Pivmecillinam hydrochloride is an analytical standard and an orally active proagent of mecillinam, an extended-spectrum penicillin antibiotic. It is intended for research and analytical applications, serving as a reference standard for assays.
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
  • Solid appearance with a white to off-white color
Catalog No. 50-004-08091