Learn More
Medchemexpress LLC Pivmecillinam hydrochloride | 32887-03-9 | MFCD01682071 | 99.9% | 476.03 g/mol | C21H34ClN3O5S | 10 MG
SDP
Catalog No. 5000298196
Encompass_Preferred
Pivmecillinam hydrochloride (standard) is supplied as a high-purity analytical reference material intended for research, method development, and assay calibration. It is the hydrochloride salt form of the prodrug pivmecillinam and is suitable for use as a reference standard in analytical laboratories.
  • High purity for analytical use (99.90%).
  • Suitable as an analytical standard and for method validation.
  • Available in small quantities for instrumental analysis, including a 10 mg size.
  • Molecular weight 476.03 g/mol and defined chemical formula for precise calculations.
Catalog No. 50-002-98196 Supplier Medchemexpress LLC Supplier No. HYB0810AR10MG
Pivmecillinam hydrochloride (standard) is supplied as a high-purity analytical reference material intended for research, method development, and assay calibration. It is the hydrochloride salt form of the prodrug pivmecillinam and is suitable for use as a reference standard in analytical laboratories.
  • High purity for analytical use (99.90%).
  • Suitable as an analytical standard and for method validation.
  • Available in small quantities for instrumental analysis, including a 10 mg size.
  • Molecular weight 476.03 g/mol and defined chemical formula for precise calculations.