Learn More
Med Vet International Povidone Iodine Scrub, 7.5%, 1 Gallon
Catalog No. 502827838
Encompass_Preferred
Microbicide Solution, Antiseptic. Helps to reduce bacteria that can potentially cause skin infection. Significantly reduces the number of microorganisms on the hands and forearms prior to surgery or patient care. Povidone Iodine 7.5% Scrub is equivalent to 0.75% titratable iodine. Surgical Hand Scrub Antiseptic Hand Wash Preoperative Skin Preparation - Compare to Betadine.
Catalog No. 50-282-7838 Supplier Med Vet International Supplier No. PHVPIVODINE
Microbicide Solution, Antiseptic. Helps to reduce bacteria that can potentially cause skin infection. Significantly reduces the number of microorganisms on the hands and forearms prior to surgery or patient care. Povidone Iodine 7.5% Scrub is equivalent to 0.75% titratable iodine. Surgical Hand Scrub Antiseptic Hand Wash Preoperative Skin Preparation - Compare to Betadine.