Learn More
Grainger Pivot Joint 63.2 mmx 40 mm 40 Series
Catalog No. 502764096
Encompass_Preferred
The 80/20 Pivot Joint is a 40mm x 40mm pivot joint made of aluminum. It allows for 360 degree rotation and can be used to connect 80/20 aluminum framing at angles. The joint has a load capacity of 560 lbs and comes with mounting hardware. It can be used in industrial, commercial, or residential settings for framing, machine building, automation, or anywhere precise adjustable angles are needed. The pivot joint enables quick and easy reconfiguration of framing and provides smooth motion.
Catalog No. 50-276-4096 Supplier Grainger Supplier No. 16U367
The 80/20 Pivot Joint is a 40mm x 40mm pivot joint made of aluminum. It allows for 360 degree rotation and can be used to connect 80/20 aluminum framing at angles. The joint has a load capacity of 560 lbs and comes with mounting hardware. It can be used in industrial, commercial, or residential settings for framing, machine building, automation, or anywhere precise adjustable angles are needed. The pivot joint enables quick and easy reconfiguration of framing and provides smooth motion.