Learn More
Aviva Systems Biology Corp PIWIL1 Antibody - N-terminal region (ARP41339_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41339_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIWIL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIWIL1. Alternate names: HIWI, PIWI, MIWIApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57809