Learn More
Medchemexpress LLC HY-13727A 5mg Medchemexpress, Pixantrone (dimaleate) CAS:144675-97-8 Purity:>98%
SDP
Catalog No. 502081482
Encompass_Preferred
Medchemexpress, HY-13727A 5mg Pixantrone (dimaleate) CAS:144675-97-8 Pixantrone dimaleate is a topoisomerase II inhibitor and DNA intercalator, with anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1482 Supplier Medchemexpress LLC Supplier No. HY13727A5MG
Medchemexpress, HY-13727A 5mg Pixantrone (dimaleate) CAS:144675-97-8 Pixantrone dimaleate is a topoisomerase II inhibitor and DNA intercalator, with anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.