Learn More
Selleck Chemical LLC Pixantrone Maleate S5059-10mg
SDP
Catalog No. 503112223
Encompass_Preferred
Pixantrone (BBR-2778) is a novel aza-anthracenedione compound with antitumor activity It is a weak topoisomerase II inhibitor and forms stable DNA adducts through alkylation with specificity for DNA hypermethylated sites
Catalog No. 50-311-2223 Supplier Selleck Chemical LLC Supplier No. S505910MG
Pixantrone (BBR-2778) is a novel aza-anthracenedione compound with antitumor activity It is a weak topoisomerase II inhibitor and forms stable DNA adducts through alkylation with specificity for DNA hypermethylated sites