Learn More
Cayman Chemical Pixantrone (maleate), Purity: ≥95%, CAS: 144675-97-8, 10 mg
SDP
Catalog No. 502982818
Encompass_Preferred
A DNA topoisomerase II inhibitor; induces SV40 DNA cleavage in the presence of mouse topoisomerase II; induces DNA single-strand breaks in NCI H187 cells; is cytotoxic to L1210 cells as well as doxorubicin-sensitive and -resistant LoVo cells (IC50 = 0.01, 0.24, and 7.2 µg/ml, respectively); decreases disease severity in a rat model of T. californica AChR immunization-induced myasthenia gravis at 8.12 mg/kg once per week for six weeks
Catalog No. 50-298-2818 Supplier Cayman Chemical Supplier No. 2005510 MG
A DNA topoisomerase II inhibitor; induces SV40 DNA cleavage in the presence of mouse topoisomerase II; induces DNA single-strand breaks in NCI H187 cells; is cytotoxic to L1210 cells as well as doxorubicin-sensitive and -resistant LoVo cells (IC50 = 0.01, 0.24, and 7.2 µg/ml, respectively); decreases disease severity in a rat model of T. californica AChR immunization-induced myasthenia gravis at 8.12 mg/kg once per week for six weeks