Learn More
Apexbio Technology LLC Pizotifen 15574-96-6 100mg
SDP
Catalog No. 502240630
Encompass_Preferred
Pizotifen (CAS 15574-96-6) is a benzocycloheptane-derived small molecule that functions as a potent antagonist of serotonin (5-HT) receptors By selectively blocking 5-HT receptors Pizotifen modulates serotonergic neurotransmission making it valuable for investigating serotonin-mediated pathways Its pharmacological actions are of interest in studies related to migraine mechanisms appetite regulation and neuropsychiatric disorders Pizotifen is frequently utilized in experimental models to elucidate the role of serotonergic signaling in both central and peripheral tissues
This item is not returnable. View return policy
Catalog No. 50-224-0630 Supplier Apexbio Technology LLC Supplier No. B1206100
Pizotifen (CAS 15574-96-6) is a benzocycloheptane-derived small molecule that functions as a potent antagonist of serotonin (5-HT) receptors By selectively blocking 5-HT receptors Pizotifen modulates serotonergic neurotransmission making it valuable for investigating serotonin-mediated pathways Its pharmacological actions are of interest in studies related to migraine mechanisms appetite regulation and neuropsychiatric disorders Pizotifen is frequently utilized in experimental models to elucidate the role of serotonergic signaling in both central and peripheral tissues