Learn More
Medchemexpress LLC Pizotifen | 15574-96-6 | MFCD00864192 | 99.6% | C19H21NS | 200 MG
SDP
Catalog No. 5000432647
Encompass_Preferred
Pizotifen, also known as Pizotyline, is a potent 5-HT2 receptor antagonist with high affinity for the 5-HT1C binding site. It is also recognized for its antidepressant properties, specifically as a 5-HT2A receptor antagonist.
  • Acts as a potent 5-HT2 receptor antagonist.
  • Shows strong affinity for the 5-HT1C binding site.
  • Functions as an antidepressant 5-HT2A receptor antagonist.
  • Capable of inhibiting serotonin-enhanced ADP-induced platelet aggregation.
  • Studies have shown no significant differences in implantations, live, dead fetuses, resorptions, or the occurrence of external, skeletal, and visceral anomalies compared to control groups.
  • No significant differences in chromosome aberrations in bone marrow cells of treated mice.
  • No elevation in micronuclei frequency in the micronucleus test.
Catalog No. 50-004-32647 Supplier Medchemexpress LLC Supplier No. HYB0115200MG
Pizotifen, also known as Pizotyline, is a potent 5-HT2 receptor antagonist with high affinity for the 5-HT1C binding site. It is also recognized for its antidepressant properties, specifically as a 5-HT2A receptor antagonist.
  • Acts as a potent 5-HT2 receptor antagonist.
  • Shows strong affinity for the 5-HT1C binding site.
  • Functions as an antidepressant 5-HT2A receptor antagonist.
  • Capable of inhibiting serotonin-enhanced ADP-induced platelet aggregation.
  • Studies have shown no significant differences in implantations, live, dead fetuses, resorptions, or the occurrence of external, skeletal, and visceral anomalies compared to control groups.
  • No significant differences in chromosome aberrations in bone marrow cells of treated mice.
  • No elevation in micronuclei frequency in the micronucleus test.